Methyl-2-[(cyanomethyl)thio]benzoate
Supplier: Apollo Scientific
OR25070-10G OR25070-1G OR25070-5G
APOSOR25070-10GEA 173 EUR
APOSOR25070-10G APOSOR25070-1G APOSOR25070-5G
Methyl-2-[(cyanomethyl)thio]benzoate
Methyl-2-[(cyanomethyl)thio]benzoate
Formula: C₁₀H₉NO₂S
MW: 207.25 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00068076
CAS Number: 57601-89-5