Tris(4-methylphenyl)phosphine 99%
Supplier: Apollo Scientific
Warning
Synonyms: Tri-p-tolylphosphine, Tris(p-tolyl)phosphine
OR25774-100G OR25774-25G OR25774-5G
APOSOR25774-100GEA 65.4 EUR
APOSOR25774-100G APOSOR25774-25G APOSOR25774-5G
Tris(4-methylphenyl)phosphine 99%
Tris(4-methylphenyl)phosphine
Formula: C₂₁H₂₁P
MW: 304.37 g/mol
Boiling Pt: 400
Melting Pt: 144…148 °C
Storage Temperature: Ambient
MDL Number: MFCD00008542
CAS Number: 1038-95-5