2,3-Dichloroquinoxaline 98%
Supplier: Apollo Scientific
Danger
OR25839-100G OR25839-25G OR25839-500G
APOSOR25839-100GEA 125 EUR
APOSOR25839-100G APOSOR25839-25G APOSOR25839-500G
2,3-Dichloroquinoxaline 98%
2,3-Dichloroquinoxaline
Formula: C₈H₄Cl₂N₂
MW: 199.04 g/mol
Boiling Pt: 301
Melting Pt: 152…154 °C
Storage Temperature: Ambient
MDL Number: MFCD00006720
CAS Number: 2213-63-0
UN: 2811
ADR: 6.1 III