4-Iodopyridine 97%
Supplier: Apollo Scientific
Warning
OR25896-1G OR25896-5G
APOSOR25896-1GEA 21.9 EUR
APOSOR25896-1G APOSOR25896-5G
4-Iodopyridine 97%
4-Iodopyridine
Formula: C₅H₄IN
MW: 205 g/mol
Melting Pt: 92…96 °C
Storage Temperature: Refrigerator
MDL Number: MFCD02093937
CAS Number: 15854-87-2