2-Chloro-6-nitroaniline 97%
Supplier: Apollo Scientific
Danger
OR26571-25G OR26571-5G OR26571-100G
APOSOR26571-25GEA 53.8 EUR
APOSOR26571-25G APOSOR26571-5G APOSOR26571-100G
2-Chloro-6-nitroaniline 97%
2-Chloro-6-nitroaniline
Formula: C₆H₅ClN₂O₂
MW: 172.57 g/mol
Boiling Pt: 80 °C/1mmHg
Melting Pt: 72…77 °C
Storage Temperature: Ambient
MDL Number: MFCD00179568
CAS Number: 769-11-9
UN: 2237
ADR: 6.1 III