5-Bromonicotinoyl chloride
Supplier: Apollo Scientific
Danger
OR26792-1G OR26792-5G
APOSOR26792-1GEA 69.6 EUR
APOSOR26792-1G APOSOR26792-5G
5-Bromonicotinoyl chloride
5-Bromonicotinoyl chloride
Formula: C₆H₃BrClNO
Melting Pt: 72…74 °C
Storage Temperature: Ambient
MDL Number: MFCD00173853
CAS Number: 39620-02-5
UN: 3261
ADR: 8,II