2,6-Dimethyl-4-hydroxybenzonitrile
Supplier: Apollo Scientific
Warning
OR26855-25G OR26855-5G OR26855-100G
APOSOR26855-25GEA 58 EUR
APOSOR26855-25G APOSOR26855-5G APOSOR26855-100G
2,6-Dimethyl-4-hydroxybenzonitrile
2,6-Dimethyl-4-hydroxybenzonitrile
Formula: C₉H₉NO
Melting Pt: 169…173 °C
Storage Temperature: Ambient
MDL Number: MFCD00238591
CAS Number: 58537-99-8
UN: 3439
ADR: 6.1,III