2-Methylpropanimidamide hydrochloride
Supplier: Apollo Scientific
Warning
Synonyms: Isobutanamidine hydrochloride, Isobutyrimidamide hydrochloride
OR27276-1G OR27276-5G OR27276-25G
APOSOR27276-1GEA 21.9 EUR
APOSOR27276-1G APOSOR27276-5G APOSOR27276-25G
2-Methylpropanimidamide hydrochloride
2-Methylpropanimidamide hydrochloride
Formula: C₄H₁₁ClN₂
Melting Pt: 159…162 °C
Storage Temperature: Ambient
MDL Number: MFCD00173787
CAS Number: 22007-68-7