2-Chloro-6-methoxyisonicotinoyl chloride
Supplier: Apollo Scientific
Danger
OR27364-1G OR27364-250MG
APOSOR27364-1GEA 60.9 EUR
APOSOR27364-1G APOSOR27364-250MG
2-Chloro-6-methoxyisonicotinoyl chloride
2-Chloro-6-methoxyisonicotinoyl chloride
Formula: C₇H₅Cl₂NO₂
Boiling Pt: 63…64 °C/0.01mmHg
Storage Temperature: Ambient
MDL Number: MFCD00125355
CAS Number: 116853-97-5
UN: 3265
ADR: 8,III