Methyl-4-aminothieno[2,3-d]pyrimidine-6-carboxylate
Supplier: Apollo Scientific
Warning
OR2757-1G OR2757-250MG
APOSOR2757-1GEA 243 EUR
APOSOR2757-1G APOSOR2757-250MG
Methyl-4-aminothieno[2,3-d]pyrimidine-6-carboxylate
Methyl-4-aminothieno[2,3-d]pyrimidine-6-carboxylate
Formula: C₈H₇N₃O₂S
MW: 209.23 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09842619
CAS Number: 155087-15-3