Tropinone
Supplier: Apollo Scientific
Warning
Synonyms: 8-Methyl-8-azabicyclo[3.2.1]octan-3-one
OR27706-25G OR27706-5G OR27706-100G
APOSOR27706-25GEA 24.8 EUR
APOSOR27706-25G APOSOR27706-5G APOSOR27706-100G
Tropinone
Tropinone
Formula: C₈H₁₃NO
MW: 139.2 g/mol
Boiling Pt: 218 °C
Melting Pt: 40…44 °C
Flash Pt: 90 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00005549
CAS Number: 532-24-1
EINECS: 208-530-6