5-(2,5-Dichlorophenyl)-2-furaldehyde
Supplier: Apollo Scientific
Warning
OR28219-1G OR28219-250MG
APOSOR28219-1GEA 102 EUR
APOSOR28219-1G APOSOR28219-250MG
5-(2,5-Dichlorophenyl)-2-furaldehyde
5-(2,5-Dichlorophenyl)-2-furaldehyde
Formula: C₁₁H₆Cl₂O₂
MW: 241.08 g/mol
Melting Pt: 97…101 °C
Storage Temperature: Ambient
MDL Number: MFCD00454841
CAS Number: 99113-89-0