2-Chloro-5-(trichloromethyl)pyridine 97%
Supplier: Apollo Scientific
Warning
OR2827-1G OR2827-5G
APOSOR2827-1GEA 21.9 EUR
APOSOR2827-1G APOSOR2827-5G
2-Chloro-5-(trichloromethyl)pyridine 97%
2-Chloro-5-(trichloromethyl)pyridine
Formula: C₆H₃Cl₄N
Melting Pt: 52…54 °C
Storage Temperature: Ambient
MDL Number: MFCD00160145
CAS Number: 69045-78-9