4-Methoxyphenylhydrazinium chloride 98%
Supplier: Apollo Scientific
Warning
OR2845-100G OR2845-25G OR2845-5G
APOSOR2845-100GEA 156 EUR
APOSOR2845-100G APOSOR2845-25G APOSOR2845-5G
4-Methoxyphenylhydrazinium chloride 98%
4-Methoxyphenylhydrazinium chloride
Formula: C₇H₁₁ClN₂O
Melting Pt: 160…162 °C (dec.)
Storage Temperature: Refrigerator
MDL Number: MFCD00012945
CAS Number: 19501-58-7
EINECS: 243-115-3
UN: 2811
ADR: 6.1,III