2-Tolualdehyde 98%
Supplier: Apollo Scientific
|
Danger
|
Synonyms:
o-Tolualdehyde, 2-Methylbenzaldehyde
OR28700-100G
OR28700-25G
OR28700-500G
APOSOR28700-100GEA
46.4
EUR
APOSOR28700-100G
APOSOR28700-25G
APOSOR28700-500G
2-Tolualdehyde 98%
2-Tolualdehyde
Formula:
C₈H₈O Boiling Pt: 199…200 °C Melting Pt: –35 °C Density: 1.039 g/cm³ (25 °C) Flash Pt: 67 °C Storage Temperature: Ambient |
MDL Number:
MFCD00003338 CAS Number: 529-20-4 EINECS: 208-452-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...