(±)-2-Methyltetrahydrofuran-3-one
Supplier: Apollo Scientific
Warning
OR28730-250G OR28730-1KG
APOSOR28730-250GEA 255 EUR
APOSOR28730-250G APOSOR28730-1KG
(±)-2-Methyltetrahydrofuran-3-one
(±)-2-Methyltetrahydrofuran-3-one
Formula: C₅H₈O₂
Boiling Pt: 139 °C
Density: 1.034 g/cm³ (25 °C)
Flash Pt: 39 °C
Storage Temperature: Ambient
MDL Number: MFCD00010423
CAS Number: 3188-00-9
EINECS: 221-685-4
UN: 1224
ADR: 3,III