2-Isopropyl-6-methylaniline
Supplier: Apollo Scientific
Warning
OR28768-5G OR28768-1G
APOSOR28768-5GEA 362 EUR
APOSOR28768-5G APOSOR28768-1G
2-Isopropyl-6-methylaniline
2-Isopropyl-6-methylaniline
Formula: C₁₀H₁₅N
MW: 149.2328 g/mol
Boiling Pt: 108…110 °C/10mmHg
Density: 0.957 g/cm³ (25 °C)
Flash Pt: 42 °C
Storage Temperature: Ambient
MDL Number: MFCD00007746
CAS Number: 5266-85-3
UN: 1993
ADR: 3 III