N-Cyclohexylaniline
Supplier: Apollo Scientific
Warning
OR28787-10G OR28787-5G
APOSOR28787-10GEA 102 EUR
APOSOR28787-10G APOSOR28787-5G
N-Cyclohexylaniline
N-Cyclohexylaniline
Formula: C₁₂H₁₇N
Boiling Pt: 279 °C
Melting Pt: 14…15 °C
Density: 0.996 g/cm³ (25 °C)
Flash Pt: 136 °C
Storage Temperature: Ambient
MDL Number: MFCD00014284
CAS Number: 1821-36-9
EINECS: 217-344-4