2-Chlorophenyl isothiocyanate 97%
Supplier: Apollo Scientific
Danger
Synonyms: 1-Chloro-2-isothiocyanatobenzene
OR28846-5G OR28846-25G
APOSOR28846-5GEA 79.9 EUR
APOSOR28846-5G APOSOR28846-25G
2-Chlorophenyl isothiocyanate 97%
2-Chlorophenyl isothiocyanate
Formula: C₇H₄ClNS
Boiling Pt: 260…262 °C
Density: 1.29 g/cm³ (25 °C)
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00004801
CAS Number: 2740-81-0
UN: 2810
ADR: 6.1,III