Dimethylthiocarbamoyl chloride
Supplier: Apollo Scientific
Danger
OR28875-100G OR28875-250G OR28875-25G
APOSOR28875-100GEA 0 EUR
APOSOR28875-100G APOSOR28875-250G APOSOR28875-25G
Dimethylthiocarbamoyl chloride
Dimethylthiocarbamoyl chloride
Formula: C₃H₆ClNS
Boiling Pt: 90…95 °C/0.5mmHg
Melting Pt: 39…43 °C
Flash Pt: 98 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00004919
CAS Number: 16420-13-6
EINECS: 240-468-5
UN: 3261
ADR: 8,II