2,3-Dichlorophenyl isothiocyanate 97%
Supplier: Apollo Scientific
Danger
Synonyms: 2,3-Dichlorophenylisothiocyanate, 1,2-Dichloro-3-isothiocyanatobenzene
OR28932-25G OR28932-5G
APOSOR28932-25GEA 221 EUR
APOSOR28932-25G APOSOR28932-5G
2,3-Dichlorophenyl isothiocyanate 97%
2,3-Dichlorophenyl isothiocyanate
Formula: C₇H₃Cl₂NS
MW: 204.08 g/mol
Boiling Pt: 256…258 °C
Density: 1.434 g/cm³ (25 °C)
Flash Pt: 113 °C
Storage Temperature: Ambient
MDL Number: MFCD00041048
CAS Number: 6590-97-2
UN: 2922
ADR: 8+6.1 II