Methyl-3-phenoxybenzoate
Supplier: Apollo Scientific
Warning
OR29298-1G OR29298-5G OR29298-10G
APOSOR29298-1GEA 24.8 EUR
APOSOR29298-1G APOSOR29298-5G APOSOR29298-10G
Methyl-3-phenoxybenzoate
Methyl 3-phenoxybenzoate
Formula: C₁₄H₁₂O₃
Storage Temperature: Ambient
MDL Number: MFCD00017192
CAS Number: 50789-43-0