2,6-Dichlorobenzaldehyde hydrazone
Supplier: Apollo Scientific
Warning
OR29357-1G OR29357-250MG OR29357-500MG
APOSOR29357-1GEA 380 EUR
APOSOR29357-1G APOSOR29357-250MG APOSOR29357-500MG
2,6-Dichlorobenzaldehyde hydrazone
2,6-Dichlorobenzaldehyde hydrazone
Formula: C₇H₆Cl₂N₂
MW: 189.05 g/mol
Melting Pt: 134 °C
Storage Temperature: Ambient
MDL Number: MFCD00729126
CAS Number: 59714-30-6