4-Morpholin-4-ylbutanenitrile
Supplier: Apollo Scientific
Warning
Synonyms: 4-Morpholinobutanenitrile
OR29418-1G OR29418-5G
APOSOR29418-1GEA 55.1 EUR
APOSOR29418-1G APOSOR29418-5G
4-Morpholin-4-ylbutanenitrile
4-Morpholin-4-ylbutanenitrile
Formula: C₈H₁₄N₂O
MW: 154.21 g/mol
Boiling Pt: 142 °C/15mmHg
Density: 1.0031 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00023393
CAS Number: 5807-11-4