4-Isopropyl-5-mercapto-4H-1,2,4-triazol-3-ol

Supplier: Apollo Scientific
Warning

OR29581-1G OR29581-5G
APOSOR29581-1GEA 95.7 EUR
APOSOR29581-1G APOSOR29581-5G
4-Isopropyl-5-mercapto-4H-1,2,4-triazol-3-ol
4-Isopropyl-5-mercapto-4H-1,2,4-triazol-3-ol
Formula: C₅H₉N₃OS
Storage Temperature: Refrigerator
MDL Number: MFCD00068125
CAS Number: 53065-47-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR