2-Cyanothioacetamide

Supplier: Apollo Scientific
Warning

OR29773-100G OR29773-25G OR29773-5G
APOSOR29773-100GEA 157 EUR
APOSOR29773-100G APOSOR29773-25G APOSOR29773-5G
2-Cyanothioacetamide
2-Cyanothioacetamide
Formula: C₃H₄N₂S
Melting Pt: 114…120 °C
Storage Temperature: Ambient
MDL Number: MFCD00010025
CAS Number: 7357-70-2
UN: 3439
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR