2-Phenoxyethanimidamide hydrochloride
Supplier: Apollo Scientific
Warning
OR29775-1G OR29775-250MG
APOSOR29775-1GEA 50.9 EUR
APOSOR29775-1G APOSOR29775-250MG
2-Phenoxyethanimidamide hydrochloride
2-Phenoxyethanimidamide hydrochloride
Formula: C₈H₁₁ClN₂O
MW: 186.64 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00173806
CAS Number: 67386-38-3