Imidazole-2-carboxylic acid ethyl ester
Supplier: Apollo Scientific
Warning
Synonyms: Ethyl imidazole-2-carboxylate, Ethyl 1H-imidazole-2-carboxylate
OR3003-5G OR3003-1G OR3003-25G
APOSOR3003-5GEA 55.1 EUR
APOSOR3003-5G APOSOR3003-1G APOSOR3003-25G
Imidazole-2-carboxylic acid ethyl ester
Imidazole-2-carboxylic acid ethyl ester
Formula: C₆H₈N₂O₂
MW: 140.14 g/mol
Boiling Pt: 48…51 °C/0.3mmHg
Melting Pt: 176…180 °C
Storage Temperature: Ambient
MDL Number: MFCD03426031
CAS Number: 33543-78-1