N'-Cyano-3-pyridinecarboximidamide
Supplier: Apollo Scientific
OR300765-1G OR300765-250MG OR300765-2.5G OR300765-5G
APOSOR300765-1GEA 249 EUR
APOSOR300765-1G APOSOR300765-250MG APOSOR300765-2.5G APOSOR300765-5G
N'-Cyano-3-pyridinecarboximidamide
N'-Cyano-3-pyridinecarboximidamide
Formula: C₇H₆N₄
MW: 146.15 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09864976
CAS Number: 73631-23-9