2-(4-Bromophenyl)ethanol
Supplier: Apollo Scientific
Warning
Synonyms: 4-Bromophenethyl alcohol
OR30084-25G OR30084-5G OR30084-100G
APOSOR30084-25GEA 79.9 EUR
APOSOR30084-25G APOSOR30084-5G APOSOR30084-100G
2-(4-Bromophenyl)ethanol
2-(4-Bromophenyl)ethanol
Formula: C₈H₉BrO
MW: 201.06 g/mol
Boiling Pt: 138 °C/9mmHg
Density: 1.436 g/cm³ (25 °C)
Flash Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD00002897
CAS Number: 4654-39-1
EINECS: 225-093-7