1-Benzyl-3-pyrrolidinone
Supplier: Apollo Scientific
Warning
OR301019-25G OR301019-5G
APOSOR301019-25GEA 46.4 EUR
APOSOR301019-25G APOSOR301019-5G
1-Benzyl-3-pyrrolidinone
1-Benzyl-3-pyrrolidinone
Formula: C₁₁H₁₃NO
Boiling Pt: 77…78 °C/0.01mmHg
Density: 1.091 g/cm³ (25 °C)
Flash Pt: 110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00005342
CAS Number: 775-16-6
EINECS: 212-274-0