tert-Butyl-4,7-diazaspiro[2.5]octane-7-carboxylate 97%
Supplier: Apollo Scientific
Warning
Synonyms: tert-Butyl 4,7-diazaspiro[2.5]octane-7-carboxylate, 4,7-Diaza-spiro(2.5)octane-7-carboxylic acid tert-butyl ester
OR301162-1G OR301162-250MG
APOSOR301162-1GEA 79.9 EUR
APOSOR301162-1G APOSOR301162-250MG
tert-Butyl-4,7-diazaspiro[2.5]octane-7-carboxylate 97%
tert-Butyl-4,7-diazaspiro[2.5]octane-7-carboxylate
Formula: C₁₁H₂₀N₂O₂
MW: 212.2887 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD08685931
CAS Number: 886766-28-5