3-Chloro-N-methylaniline
Supplier: Apollo Scientific
Warning
OR30117-1G OR30117-25G OR30117-5G
APOSOR30117-1GEA 0 EUR
APOSOR30117-1G APOSOR30117-25G APOSOR30117-5G
3-Chloro-N-methylaniline
3-Chloro-N-methylaniline
Formula: C₇H₈ClN
Boiling Pt: 243…244 °C
Density: 1.156 g/cm³ (25 °C)
Flash Pt: >110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00052014
CAS Number: 7006-52-2