6-Hydroxy-2-azaspiro[3.3]heptane-2-carboxylic acid tert-butyl ester
Supplier: Apollo Scientific
Warning
Synonyms: 6-Hydroxy-2-azaspiro[3.3]heptane, N-BOC protected, tert-Butyl 6-hydroxy-2-azaspiro[3.3]heptane-2-carboxylate
OR301181-1G OR301181-250MG
APOSOR301181-1GEA 24.8 EUR
APOSOR301181-1G APOSOR301181-250MG
6-Hydroxy-2-azaspiro[3.3]heptane-2-carboxylic acid tert-butyl ester
6-Hydroxy-2-azaspiro[3.3]heptane-2-carboxylic acid tert-butyl ester
Formula: C₁₁H₁₉NO₃
MW: 213.28 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD15071431
CAS Number: 1147557-97-8