1-Methylpiperazin-2-one 95%
Supplier: Apollo Scientific
Warning
OR301209-1G OR301209-5G OR301209-250MG
APOSOR301209-1GEA 65.4 EUR
APOSOR301209-1G APOSOR301209-5G APOSOR301209-250MG
1-Methylpiperazin-2-one 95%
1-Methylpiperazin-2-one
Formula: C₅H₁₀N₂O
MW: 114.1457 g/mol
Boiling Pt: 134…137 °C/0.5mmHg
Storage Temperature: Ambient
MDL Number: MFCD00102572
CAS Number: 59702-07-7