4-Chlorophenyl isothiocyanate 98%
Supplier: Apollo Scientific
Danger
OR30198-25G OR30198-100G
APOSOR30198-25GEA 109 EUR
APOSOR30198-25G APOSOR30198-100G
4-Chlorophenyl isothiocyanate 98%
4-Chlorophenyl isothiocyanate
Formula: C₇H₄ClNS
Boiling Pt: 135…136 °C/24mmHg
Melting Pt: 42…46 °C
Flash Pt: >113 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00004810
CAS Number: 2131-55-7
EINECS: 218-358-3
UN: 2923
ADR: 8+6.1,III