2-(Boc-amino)-4-bromopyridine
Supplier: Apollo Scientific
Warning
Synonyms: tert-Butyl (4-bromopyridin-2-yl)carbamate
OR302007-1G OR302007-5G
APOSOR302007-1GEA 82.8 EUR
APOSOR302007-1G APOSOR302007-5G
2-(Boc-amino)-4-bromopyridine
2-(Boc-amino)-4-bromopyridine
Formula: C₁₀H₁₃BrN₂O₂
MW: 273.13 g/mol
Melting Pt: 139…140 °C
Storage Temperature: Ambient
MDL Number: MFCD09037866
CAS Number: 207799-10-8