2-Indolinone
Supplier: Apollo Scientific
Warning
Synonyms: 2-Oxindole, Oxindole, Indolin-2-one, 2,3-Dihydro-2-indolone
OR30278-25G OR30278-5G
APOSOR30278-25GEA 24.8 EUR
APOSOR30278-25G APOSOR30278-5G
2-Indolinone
2-Indolinone
Formula: C₈H₇NO
MW: 133.15 g/mol
Boiling Pt: 227 °C/73mmHg
Melting Pt: 123…128 °C
Storage Temperature: Ambient
MDL Number: MFCD00005711
CAS Number: 59-48-3
EINECS: 200-429-5