2,2-Dimethylaziridine
Supplier: Apollo Scientific
Warning
OR303021-1G OR303021-500MG
APOSOR303021-1GEA 538 EUR
APOSOR303021-1G APOSOR303021-500MG
2,2-Dimethylaziridine
2,2-Dimethylaziridine
Formula: C₄H₉N
MW: 71.12 g/mol
Boiling Pt: 70…72 °C
Storage Temperature: Ambient
MDL Number: MFCD00040452
CAS Number: 2658-24-4
EINECS: 220-191-6