Isopropyl isothiocyanate
Supplier: Apollo Scientific
Danger
Synonyms: Isopropylisothiocyanate
OR30306-100G OR30306-25G
APOSOR30306-100GEA 143 EUR
APOSOR30306-100G APOSOR30306-25G
Isopropyl isothiocyanate
Isopropyl isothiocyanate
Formula: C₄H₇NS
MW: 101.17 g/mol
Boiling Pt: 136…138 °C
Density: 0.948 g/cm³ (25 °C)
Flash Pt: 40 °C
Storage Temperature: Ambient
MDL Number: MFCD00037830
CAS Number: 2253-73-8
EINECS: 218-851-3
UN: 1992
ADR: 3+6.1 II