N,2,6-Trimethylaniline
Supplier: Apollo Scientific
Warning
OR303237-1G OR303237-500MG
APOSOR303237-1GEA 538 EUR
APOSOR303237-1G APOSOR303237-500MG
N,2,6-Trimethylaniline
N,2,6-Trimethylaniline
Formula: C₉H₁₃N
MW: 135.21 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00613418
CAS Number: 767-71-5