6-(Cyanomethyl)nicotinonitrile
Supplier: Apollo Scientific
Warning
OR303677-1G OR303677-500MG
APOSOR303677-1GEA 400 EUR
APOSOR303677-1G APOSOR303677-500MG
6-(Cyanomethyl)nicotinonitrile
6-(Cyanomethyl)nicotinonitrile
Formula: C₈H₅N₃
MW: 143.15 g/mol
Melting Pt: 78…80 °C
Storage Temperature: Ambient
MDL Number: MFCD09924285
CAS Number: 1000516-33-5