2-Chloro-1-(4-hydroxy-3-methoxyphenyl)propan-1-one 97%
Supplier: Apollo Scientific
OR303967-1G OR303967-500MG OR303967-5G
APOSOR303967-1GEA 336 EUR
APOSOR303967-1G APOSOR303967-500MG APOSOR303967-5G
2-Chloro-1-(4-hydroxy-3-methoxyphenyl)propan-1-one 97%
2-Chloro-1-(4-hydroxy-3-methoxyphenyl)propan-1-one
Formula: C₁₀H₁₁ClO₃
Melting Pt: 77…78 °C
Storage Temperature: Ambient
MDL Number: MFCD22373656
CAS Number: 68505-86-2