(S)-(+)-Hydratropic acid 97+%
Supplier: Apollo Scientific
Warning
Synonyms: (S)-2-Phenylpropanoic acid, (S)-(+)-2-Phenylpropionic acid, (S)-(+)-2-Phenylpropanoic acid
OR304086-100G OR304086-25G OR304086-5G
APOSOR304086-100GEA 724 EUR
APOSOR304086-100G APOSOR304086-25G APOSOR304086-5G
(S)-(+)-Hydratropic acid 97+%
(S)-(+)-Hydratropic acid
Formula: C₉H₁₀O₂
MW: 150.18 g/mol
Boiling Pt: 115 °C/0.75mmHg
Melting Pt: 29…30 °C
Density: 1.1 g/cm³ (25 °C)
Flash Pt: 113 °C
Storage Temperature: Ambient
MDL Number: MFCD00063139
CAS Number: 7782-24-3