Methyl-3-aminothiophene-2-carboxylate
Supplier: Apollo Scientific
Warning
Synonyms: Methyl 3-amino-2-thiophenecarboxylate
OR30429-100G OR30429-25G
APOSOR30429-100GEA 24.8 EUR
APOSOR30429-100G APOSOR30429-25G
Methyl-3-aminothiophene-2-carboxylate
Methyl 3-aminothiophene-2-carboxylate
Formula: C₆H₇NO₂S
Boiling Pt: 100…102 °C/0.1mmHg
Melting Pt: 62…64 °C
Storage Temperature: Ambient
MDL Number: MFCD00009765
CAS Number: 22288-78-4
EINECS: 244-895-8