trans-Cyclobutane-1,2-dicarbonitrile
Supplier: Apollo Scientific
OR304349-1G OR304349-5G
APOSOR304349-1GEA 209 EUR
APOSOR304349-1G APOSOR304349-5G
trans-Cyclobutane-1,2-dicarbonitrile
trans-Cyclobutane-1,2-dicarbonitrile
Formula: C₆H₆N₂
Storage Temperature: Ambient
MDL Number: MFCD00221717
CAS Number: 3211-20-9