4-Chloro-3-ethylphenol 98%
Supplier: Apollo Scientific
Warning
OR30478-25G OR30478-5G
APOSOR30478-25GEA 92.8 EUR
APOSOR30478-25G APOSOR30478-5G
4-Chloro-3-ethylphenol 98%
4-Chloro-3-ethylphenol
Formula: C₈H₉ClO
Melting Pt: 45…50 °C
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00010822
CAS Number: 14143-32-9
EINECS: 237-991-6