tert-Butyl-4-(4-aminobenzyl)piperazine-1-carboxylate
Supplier: Apollo Scientific
Warning
Synonyms: 4-(4-Aminobenzyl)piperazine, N1-BOC protected, tert-Butyl 4-(4-aminobenzyl)piperazine-1-carboxylate
OR305321-1G OR305321-5G
APOSOR305321-1GEA 34.8 EUR
APOSOR305321-1G APOSOR305321-5G
tert-Butyl-4-(4-aminobenzyl)piperazine-1-carboxylate
tert-Butyl-4-(4-aminobenzyl)piperazine-1-carboxylate
Formula: C₁₆H₂₅N₃O₂
MW: 291.3886 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06797795
CAS Number: 304897-49-2