Octachloronaphthalene 98%
Supplier: Apollo Scientific
|
Warning
|
OR30671-1G
OR30671-25G
OR30671-5G
APOSOR30671-1GEA
29
EUR
APOSOR30671-1G
APOSOR30671-25G
APOSOR30671-5G
Octachloronaphthalene 98%
Octachloronaphthalene
Formula:
C₁₀Cl₈ Boiling Pt: 440 °C Melting Pt: 194…197 °C Density: 2 g/cm³ (25 °C) Flash Pt: >110 °C Storage Temperature: Ambient |
CAS Number:
2234-13-1 EINECS: 218-778-7 |