2,6,8-Trimethyl-4-quinolinol
Supplier: Apollo Scientific
Synonyms: 4-Hydroxy-2,6,8-trimethylquinoline
OR307882-1G OR307882-5G
APOSOR307882-1GEA 293 EUR
APOSOR307882-1G APOSOR307882-5G
2,6,8-Trimethyl-4-quinolinol
2,6,8-Trimethyl-4-quinolinol
Formula: C₁₂H₁₃NO
Storage Temperature: Ambient
CAS Number: 15644-93-6